Indoors, air quality can be monitored for hazardous levels. In schools, office buildings, greenhouses, and anywhere that high levels of carbon dioxide are generated, the Extech Portable Indoor Air Quality CO2 Meter is a good option for checking air quality. For personal monitoring badges, the TraceAir® Personal Organic Solvent Panel/Indoor Air Quality Badge is a high sampling rate monitor to use when organic vapors are suspected to be the cause of indoor air quality concerns. The Organic Solvent Panel (OSP) badge includes analysis by GC/FID for 25 common organic vapors.
To clean air, Cloud 9® Air Cleaner with HEPA-Filter is EPA-registered and traps particles to 0.3µm. To rid your lab, office, or waiting room of dust, pollen, viruses, bacteria, mold spores, and smoke particles with a portable air cleaner, choose the Compact Air Cleaner with HEPA-Filter and Optional VOC Filter. This unit even removes gases associated with sick building syndrome.
Fantastic write-up, I am a large believer in placing comments about web sites to inform your blog copy writers realize that theya’ve added some thing of great benefit around the globe wide internet!
Thanks for your response, Marry.