Affordable Benchtop pH Meters with Lab-Friendly Features? Watch the Video!
Oakton® 2700 Series Benchtop pH Meters have a lot going for them: an oversized display with bright backlight, high 0.001 pH resolution with ±0.002 pH accuracy, and store and recall function for up to 500 sets of data. Yet, they remain affordable. And, they meet GLP requirements, stamping all calibration and stored data with time/date.
Take a look to see how these meters might enhance your lab:
Be the first to comment on "Affordable Benchtop pH Meters with Lab-Friendly Features? Watch the Video!"