Oakton Economy pH Meter featured on CBS’s CSI Miami
pH meter
The Oakton economy pH meter hits prime time.
See it on CBS’s CSI Miami – Episode 24 “Dissolved”. The show featured the Oakton® economy pH meter and a probe and wa
The microprocessor-based Oakton Economy pH 11 Meter takes measurements in pH, mV or °C. Use mV output to take ORP readings. Other features include
  • Hold function, automatic shutoff, and ready indicator.
  • Dual display of pH (or mV) and temperature.
  • Push-button pH calibration at up to five points and offset mV calibration
  • Data storage for up to 50 points
See the complete line up of Oakton products.
Oakton provides incredible quality at budget-friendly prices. Their selection of water quality instruments includes pH, ion, conductivity, and dissolved oxygen meters.
Be the first to comment on "Oakton Economy pH Meter featured on CBS’s CSI Miami"