Using a Versatile Gas Mass Flow Controller
Cole-Parmer® Flow Gas Mass Flow Controller, for gas, 0.1 to 10 LPM
Cole-Parmer® Flow Gas Mass Flow Controller, for gas, 0.1 to 10 LPM
What happens when a customer really likes an instrument? A professional researcher used the Cole-Parmer® Flow Gas Mass Flow Controller for Gas and wrote to us:
“These are very flexible instruments, pre-loaded with calibration data for a wide range of gases so they can be switched to a different gas as needed. This makes them much more versatile than comparably priced controllers that are calibrated for only one gas. They also report several condition parameters (outlet pressure, temperature, either volumetric or mass flow) that can be logged during an experiment. This feature also makes them more generally useful than other comparably priced controllers.”
Because the unit worked so well for this professional, he recommends it for use in labs and research.
Be the first to comment on "Using a Versatile Gas Mass Flow Controller"