Easier Small Volume Distillations
SimpleDist® Micro
SimpleDist® Micro
Smaller samples save time. The new Environmental Express® SimpleDist® Micro distills 26 samples simultaneously for easier distillation. It also eliminates the need for costly glassware through the use of disposable polypropylene SimpleDist tubes. No cooling water is required.
The EPA has approved the use of micro distillation systems with disposable tubes for reporting on cyanide. The system can also adapt to report on ammonia and phenols as well.
The SimpleDist® Micro distillation system follows established EPA methods 335.4 and 350.1, and Standard Methods 4500-CN-C and 4500-NH3.
With proven HotBlock® dependability, the SimpleDist Micro heating block is made from PTFE-coated graphite enclosed in a durable Kydex® housing with a built-in controller.
Need more information or have questions? Contact out technical support team.
Be the first to comment on "Easier Small Volume Distillations"