Make your manufacturing, construction, or mining environment less noisy by using an air operated pump. An internal muffler reduces air noise while the pump itself consumes 60% less air. Less noise means fewer distractions and greater ability to concentrate on the task in front of you.
Air Operated Pumps handle petroleum-based lubricants and synthetic oils and can be used for any application requiring pressurized oil. They are rugged enough for even extreme environments.
The pump’s stall-free air motor can operate with or without a lubricator. You can easily service the pump in-line to minimize downtime. You can also make quick orientation changes by adjusting the outlet up to 360°. Pumps can be mounted directly to a drum or tank.
Be the first to comment on "Decrease the Noise with Air-Operated Pumps"