PCRMax Alpha Cyclers: Thermal Cyclers for Simplified Optimization – Watch Video
Would you like to optimize your assays? easily create, modify and access programs, saving time? Look at these compact thermal cyclers that deliver reproducible results every run—with little to no training needed. The PCRmax Alpha Cyclers are developed to deliver not only the best quality data you can expect from PCRmax but also to innovate and exceed the high standards expected by the community.
The PCRmax Alpha Thermal Cyclers are developed to deliver not only the best quality data you can expect from PCRmax but also to innovate and exceed the high standards expected by the community.
Alpha Cyclers’ software has features such as recently used programs; allowing you to quickly access your most commonly used protocols without the need to navigate through folders to find it, individual user logins; with protected protocols as-well-as allowing users’ access to temperature logs after each run to monitor the state of the system.
The Alpha Cycler systems also contain a novel program wizard which allows you to define a protocol based specifically off their primer sequence and template source. Primer sequences or Tms are inputted, amplicon length and source defined and the on board algorithm generates an optimized protocol for your assay.
  • Scalable – Chose from any combination of 96 or 384 well blocks
  • Compact – Save precious lab space by condensing four truly independent blocks into one chassis with no networking issues or connectivity concerns
  • Active Sample cooling – For sharper amplification and minimal non-specific amplification
  • Programme Wizard – Generate a protocol specific to your sequence, template source and amplicon length in seconds
  • Gradient – Allowing for simplified temperature optimization, no matter the block chosen
  • Report generated on run conditions and state following completion of protocol
  • 96/384 well formats – Flexibility for any scale and user needs USB connectivity – To retrieve system information and easily transfer protocols between systems
  • Android driven 10” tablet interface – Quad core speed and excellent connectivity and feel
Need more information on the PCRmax Alpha Cyclers? Contact our technical experts at 1-800-323-4340.
Be the first to comment on "PCRMax Alpha Cyclers: Thermal Cyclers for Simplified Optimization – Watch Video"