Oakton® EcoTestr™pH2+ and CTS pocket meters are winners
The improved Oakton EcoTestr pH2+ and CTS pocket testers provide more user-friendly options to make water analysis easier and faster—no wonder editors at Pool & Spa News (PSN) selected the Oakton EcoTestr as one of their 2018 Top 50 Products! According to PSN, the chosen products are the result of reader response to the products in the PSN 2018 New Products issue and the editor’s consideration of a vast selection of products.
Here’s why these testers should be in your pocket:
The Oakton EcoTestr pH2+ Pocket Meter has an expanded 4-digit dual-line display shows pH and temperature simultaneously. The meter also has up to three-point calibration. It is ideal for pools & spas, education, aquariums, and anywhere else pH is measured.
The Oakton EcoTestr CTS Pocket Meter is now a 3-in-1 tool that can measure conductivity, total dissolved solids and salinity. The expanded 4-digit dual-line display shows the parameter and temperature simultaneously. The meter also has single-point calibration. This meter is suited for water treatment, environmental monitoring, and anywhere conductivity, TDS and salinity are monitored.
Both the pH2+ and the CTS pocket testers have a larger display with a better viewing angle and a more intuitive interface. To enhance field use, the improved cap is now leakproof and functions as a sample cup and as a stabilizing base to stand the meter upright. With an IP67-rated housing, the meters are dustproof and waterproof—and they float. A lanyard attachment makes field measurements more convenient.
Browse our Oakton 50 Series Pocket Meters
See our complete line of pocket testers
Browse the complete line of Oakton products
Be the first to comment on "Oakton EcoTestr: Pool & Spa News 2018 Top 50 Product"