Hundreds of products are on sale now!
We are celebrating the 4th of July by giving you savings on hundreds of items. For a limited time, save up to 50% on many customer favorites. Here is a peek at some of the products you can find on sale during our July 4th Salebration:
Argos Technologies® Omega Pipette Controllers
These compact pipette controllers feature an ergonomic rounded design for comfortable hand fit and light-action button springs for minimal activation pressure. The UV-resistant plastic housing is lightly textured to ensure a slip-resistant grip and is available in six colors. Five-bar LED battery charge indicator lights up during use; each bar represents 20% of the rechargeable lithium batteries charge capacity.
Cole-Parmer® StableTemp™ -86ºC Upright ULT Freezers
Designed to meet your cold storage needs down to -86ºC, these ULT freezers are built with temperature stability and energy savings in mind each is designed for optimal efficiency with minimal temperature loss. The cabinet consists of high-performance insulation panels that minimize heat gain improving temperature stability. Separate inner storage chambers have their own insulated doors which provide independent access to storage space, minimizing frost buildup inside the chambers and maximum protection of stored samples. Outer door seal’s unique design reduces temperature loss while the door is open, and a robust door latch keeps the door securely closed.
Oakton® EcoTestr™ pH1 Waterproof Pocket Tester
Need a water tester? This economical, simple-to-use pH tester measures pH levels and temperatures quickly, displaying them both simultaneously on a large dual-line LCD. It has a pH range from 0 to 14 with an accuracy of ±0.1 pH. The tester features up to three pH calibration points with auto NIST and US buffer recognition. Additional features include a long-lasting durable electrode with polymer reference electrolyte; status icons for measurement stability, calibration, and battery life; and self-diagnostics. The IP67-rated housing features an easy-to-grip design and floats if accidentally dropped in water when sensor cap is on.
Cole-Parmer® MS-3400 Centrifuges
Be the first to comment on "Saving are Exploding this 4th of July"