No Image
What Not To Do With Ultrasonic Cleaners
Ultrasonic Cleaners use cavitation, created by ultrasonic waves, to scrub objects made of nonabsorbent materials in the research, pharmaceutical, printing, electronics, engineering, and automotive industries. Higher frequency models clean more gently, precisely, and reliably. Some…