During Balance Awareness Week, An Overview of Maintaining a Correct Balance
Let’s start with the basics. A balance measures the mass of an item by comparing the mass of a known object to an unknown object. In electrical balances, a load cell containing a strain gauge…