sample prep
How To Select Homogenizers to Enhance Tissue Processing
Homogenizing tissue samples quickly and efficiently. When choosing homogenizers, which homogenizer do you choose when you need to homogenize tissue samples quickly and efficiently, without cross-contamination? Consider your application, sample size, speed needs, probes, and…
microscope optics
What are infinity optics on a microscope?
Microscopes with infinity corrected optical systems have infinity image distance. These systems are designed by placing a tube lens within the body tube between the objective and the eyepieces, resulting in an intermediate image. This…