Summer Fun Begins with Proper pH
A comprehensive selection of Oakton pH testers, conductivity meters, and calibration solutions, ideal for precise and reliable measurements in laboratory and field settings.
Summer is the perfect time to enjoy your pool, but maintaining the right pH levels is essential for ensuring a safe and enjoyable swimming experience. Here’s everything you need to know about keeping your pool’s pH in check.
Oakton 35634-30 Waterproof Pocket pH Tester
Oakton 35634-30 Waterproof Pocket pH Tester
What is pH and Why is it Important?
pH measures the acidity or alkalinity of your pool water on a scale from 0 to 14. A pH level of 7 is neutral, while levels below 7 are acidic, and those above are alkaline. Maintaining a balanced pH is crucial for several reasons:
  • Comfort:
    Proper pH levels prevent skin and eye irritation, making swimming more enjoyable.
  • Safety:
    A balanced pH ensures the effectiveness of chlorine and other sanitizers, keeping harmful bacteria at bay.
  • Equipment Longevity:
    Improper pH can lead to corrosion or scaling, damaging your pool’s equipment and surfaces.
Ideal pH Levels for Pools
For optimal swimming conditions, aim for a pH level between 7.2 and 7.8. This range ensures comfort and maximizes the efficiency of your pool’s chlorine. Regular testing is key to maintaining this balance.
How to Test Your Pool’s pH
A comprehensive selection of Oakton pH testers, conductivity meters, and calibration solutions, ideal for precise and reliable measurements in laboratory and field settings.
Testing your pool’s pH is simple and can be done using a variety of methods:
  • Test Strips:
    Dip a test strip into the pool water and compare the color to a chart.
  • Digital Testers:
    These provide accurate readings and are easy to use.
  • Liquid Test Kits:
    Add pool water to a tube, add the reagent, and compare the color change to a chart.
It’s recommended to test your pool’s pH at least twice a week, especially during heavy use or after rainstorms.
Adjusting Your Pool’s pH
If your pool’s pH is too high or too low, you can adjust it using common pool chemicals:
  1. To Raise pH: Add sodium carbonate (soda ash).
  2. To Lower pH: Add muriatic acid or sodium bisulfate.
Always follow the manufacturer’s instructions and safety guidelines when handling pool chemicals.
Recommended Products:
  • Oakton pH 700 Benchtop Meter: Ideal for ensuring accurate pH readings, this meter offers precision and ease of use. Electrode sensors include a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage. The keypad clicks to confirm operation and allows access to a variety of functions. Calibrate up to three pH points with auto-buffer recognition at both USA and NIST standard buffers.
  • Oakton EcoTestr pH 2+ Pocket Meter: A portable solution for quick and reliable pH testing on the go. Additional features include automatic temperature compensation (ATC), a hold function, an auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
  • Oakton pH Tester 1 Pocket Meter: A portable low cost, accurate solution for quick and reliable pH testing on the go. Additional features include automatic temperature compensation (ATC), 3-point calibration, and 0.1 accuracy.
  • Oakton PCTS 5 Tester pH/EC/TDS/Temperature: This multifunctional tester provides comprehensive water quality analysis.
Maintaining pH Balance
To keep your pool’s pH balanced, consider the following tips:
  1. Monitor Alkalinity:
    Total alkalinity acts as a buffer for pH, helping to stabilize it. Keep it between 80-120 ppm.
  2. Regular Maintenance:
    Clean your pool and filter regularly to prevent debris from affecting the water chemistry.
  3. Consistent Testing:
    Regular testing allows you to catch and correct imbalances quickly.
Enjoy a Safe and Refreshing Summer
By maintaining the proper pH levels, you’ll ensure that your pool remains a safe and inviting place for summer fun. With a little attention and regular maintenance, you can enjoy crystal-clear water all season long. Dive in and make the most of your summer!
Explore our entire catalog of water quality products today!