Introducing the picoSpin™ Benchtop NMR Spectrometer from Cole-Parmer―a true breakthrough in chemical instrumentation! Researchers and educators now have access to a revolutionary benchtop NMR spectrometer.
The 45-MHz picoSpin spectrometer resolves proton chemical shifts with a resolution of 100 parts per billion using a very small sample volume of less than 20 microliters. Liquid samples are simply injected into an internal capillary via front-panel fittings.
“We have engineered an innovative and disruptive technology that changes the NMR spectroscopy landscape. With its low price, compact footprint and true spectroscopic capability, the picoSpin-45 spectrometer enables applications that could not be considered in the past,” said John Price, Ph.D., president and chief scientist of picoSpin, LLC.
The complete liquid-phase proton NMR system includes permanent magnet, transmitter, receiver, data acquisition, programmable pulse sequencer, Ethernet interface, and intuitive web-based control software.
Be the first to comment on "View the world’s first miniature NMR spectrometer―affordable, portable, and easy to use"