Thermo Scientific Nalgene Narrow-Mouth Clear Polycarbonate Bottles
Safely store samples at sub-zero tempearture
  • Polypropylene screw cap prevents leakage
  • Autoclavable
These clear polycarbonate (PC) bottles can be stored at sub-zero temperatures. They have leakproof polypropylene (PP) screw closures. Four- and eight-liter sizes have a built-in shoulder loop. Replacement caps sold separately.
$52.30 - $176.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene DS2205-0210 Polycarbonate Narrow-Mouth Bottle,  2 L
Buy More Save More
EW-06063-50 Mfr # DS2205-0210
COMPARE
2 38-430 Polycarbonate(PC)
$52.30USD / Each
Thermo Scientific Nalgene DS2205-0250 Polycarbonate Narrow-Mouth Bottle,  2.5 L
Buy More Save More
EW-06063-52 Mfr # DS2205-0250
COMPARE
2.5 38-430 Polycarbonate(PC)
$60.00USD / Each
Thermo Scientific Nalgene DS2205-0010 Polycarbonate Narrow-Mouth Bottle,  4 L
Buy More Save More
EW-06063-54 Mfr # DS2205-0010
COMPARE
4 38-430 Polycarbonate(PC)
$107.00USD / Each
Thermo Scientific Nalgene DS2205-0020 Polycarbonate Narrow-Mouth Bottle,  8 L
Buy More Save More
EW-06063-56 Mfr # DS2205-0020
COMPARE
8 53B Polycarbonate(PC)
$176.00USD / Each