Environmental Express Oakton pHTestr® 30+ Waterproof Pocket Tester

Representative image only.

Environmental Express Oakton pHTestr® 30+ Waterproof Pocket Tester

Environmental Express Oakton | Item: UX-35634-30

Durable pocket meters for the most challenging routine applications

$164.35USD / Each

for an additional
$104.50USD / Each
for an additional
$187.50USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$164.35USD / Each
Product Description

Includes

4 AAA batteries.

Key Features

  • Replaceable, double-junction electrode sensors for highly caustic, volatile or hazardous samples
  • Easy to operate Three-button system
  • IP67-rated housing sits flat on the table and floats in water
  • 0.01 pH accuracy along with dual pH and selectable C or F temperature display
  • IP67-rated housing sits flat on the table and floats in water
  • Optional sensors for ORP testing available (single or double-junction)
  • 1 year warranty

More About this Item

Our most capable series of pocket testers, the Oakton 30 Series include pHTestr 30, ORPTestr 30, and PCTSTestr 30. Replaceable and double junction sensors combine to provide durability and longevity against challenging substance properties in both lab and field applications. Waterproof and designed to float in water, they are ideal for field-based testing environments – with a convenient loop for security line attachment.

30 series pocket meters include a replaceable double-junction electrode sensor that is designed for more aggressive applications, including wastewater and solutions containing sulfides, heavy metals, and TRIS. The electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage and allow for easy maintenance and replacement. A Valox sensor housing resists most chemicals.

An easy-to-use three-button system allows for technician level use and measurement. Up to 3 calibration points with auto-buffer recognition at both USA and NIST standard buffers. Additional features include an automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.

Waterproof pHTestr 30 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can attach a line to the housing for easy retrieval. Additionally, the Valox sensor housing resists most chemicals.

The pHTestr 30 has a replaceable double-junction electrode sensor that is designed for more aggressive applications, including wastewater and solutions containing sulfides, heavy metals, and TRIS. The electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage and allow for easy maintenance. A Valox sensor housing resists most chemicals.

An easy-to-use three-button keypad allows for quick access to a variety of functions. Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers. Additional features include an automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.

pHTestr 30 purchased before Oct. 2019 use replacement sensor 35624-38. pHTestr 30 purchased after Oct. 2019 use replacement sensor 35634-03.

Meter includes: protective plastic storage case, lanyard, and batteries.

IP 671 Year WarrantyISO9001:2000 Certified Supplier
Specifications
  • Meter TypeTester
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01
  • pH Accuracy±0.01
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • Min Temperature (° F)32
  • Max Temperature (° F)122
  • Temperature Resolution0.1
  • Temperature Accuracy0.9 F, 0.5 C
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 AAA (included)
  • Battery life (hr)500
  • Diameter (cm)3.81
  • Diameter (in)1 1/2
  • DescriptionWaterproof pH Testr 30 Pocket Tester
  • Warranty1 year
Reviews
Q&A