DescriptionTissue Embedding Station 120 VAC 50/60Hz
Warranty1 Year
Key Features
Programmable start times ensure dependability and performance
User-friendly membrane touch pad allows for precise control
Cold plate offers compressor-cooled work surface
1 year warranty
More About this Item
This tissue embedding station requires less bench space than most in its class by combining various work zones into one functional unit. The comfortable armrest minimizes fatigue and reduces heat — ideal for moderate to heavy workloads. The station comes equipped with a microprocessor-based controller and a user-programmable timer to provide accurate temperature regulation and convenient monitoring of independently controlled heating and cooling elements.
Station features convenient zones that can be programmed to cycle on/off or remain on continuously. The zones include a paraffin reservoir, warming oven, tissue holding tank, illuminated work stage, and compressor-cooled cold plate. The large 3.75-liter (1-gallon) paraffin reservoir reduces the need for constant refilling and allows for adjustment of flow rate from 0 to 36 mL/min. The unique self-draining system eliminates excess paraffin—keeping the lab safe and clean. The spacious warming oven with easy-access hinged lid can accommodate various sizes of base molds.
The tissue holding tank, complete with drainage shelf and debris screen, accepts various tissue processor baskets. Easily dispense paraffin wax using the infrared-activated dispenser or the footswitch, eliminating the need for hand contact with problematic mechanical switches. The Freon-chilled cold spot is ideal for high volume workloads and maintains lower temperatures than systems that use Peltier heat exchangers.
The system features built-in support for optional accessories including a dual-power magnifying glass and electronic forceps. The electronically heated forceps plug right into the embedding center and are useful for smoothing wrinkled tissue and paraffin samples. Use the adjustable magnifying glass to facilitate specimen orientation. Order accessories separately.