This item is now discontinued. We recommend the following as suggested replacements: EW-30009-14
Digi-Sense Video Borescope, 320 x 240 Resolution
Representative image only.
Digi-Sense Item # EW-20250-27
Ideal for inspecting duct work, wiring locations, piping, vehicles, and much more
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionVideo Borescope, 320 x 240 Resolution
  • Warranty2 year
Two year warrantyConformite EuropeenneIP 67
Key Features
  • 2 year warranty
More About this Item
Flexible camera-tipped probes provide easy viewing in hard-to-reach areas. Bright LCD monitor allows instant viewing of area being inspected. Adjustable LED light on camera tip provides the necessary light in dimly lit areas or adjusts down when viewing reflective objects to eliminate washout.
Digi-Sense Video Borescope 150
* Capture both still and video images for playback on LCD, TV, or computer monitor
* Transfer video and images to your PC with SD card or USB cable
* Images can be captured with date/time stamp
* 8 GB flash memory
* 3.2" color LCD monitor for real-time viewing
* TV-Out hookup for real-time or recorded viewing on external video devices
* Four adjustable LED lights for operation in dark environments
* Camera probe is IP67 rated (waterproof) for wet environments
* 180° image rotation on LCD
* Built-in Li battery with USB charging
* Languages: English, French, German, and Spanish