Digi-Sense Type K High-Temperature Flexible Thermocouple Probe; 12" L
Representative image only.
Digi-Sense Type K High-Temperature Flexible Thermocouple Probe; 12" L
Digi-Sense | Item: EW-08516-96
High-Temperature Thermocouple Probes Designed for Multiple Bends Ideal for any Environment
$66.60USD / Each
BUY MORE & SAVE
  • 1-5$66.60 USD / Each
  • 6-11$59.94 USD / Each
  • 12-23$56.61 USD / Each
  • 24+$53.28 USD / Each
for an additional
$231.00USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$66.60USD / Each
BUY MORE & SAVE
  • 1-5$66.60 USD / Each
  • 6-11$59.94 USD / Each
  • 12-23$56.61 USD / Each
  • 24+$53.28 USD / Each
Buy It Together
Current Item
Digi-Sense Type K High-Temperature Flexible Thermocouple Probe; 12" L
Digi-Sense Type K High-Temperature Flexible Thermocouple Probe; 12" L
0851696
$66.60USD / Each
+ + + =
Total Price 1686.28 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Bend probes around pipe diameters as small as twice the sheath diameter to easily determine your pipe temperature
  • Ideal for industrial environments where fast accurate readings are required
  • Response Time: .85 seconds or greater
  • Type “K” and “J” probes feature ICONEL 600 sheaths
  • Type “T” probes feature 316 Stainless Steel sheaths
  • Fiberglass insulated cable with stainless steel overbraid
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty
More About this Item
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
6 Month Warranty
Specifications
  • Probe TypeK
  • Length (")12
  • Min Temperature (° F)-418
  • Max Temperature (° F)2012
  • Min Temperature (° C)-250
  • Max Temperature (° C)1100
  • JunctionGrounded
  • Connector TypeMini Connector
  • Sheath Length (in)12
  • DescriptionType K High-Temperature Flexible Thermocouple Probe; 12" L