Help
Still have questions?
Need help? Contact us for technical assistance.
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 30 pages of laboratory, industrial, and in-line mixers, including accessories, from such names as Cole-Parmer, IKA, Lightnin, and Heidolph. Request yours today.
Featured Products
IKA RW20 Overhead Lab Mixer
IKA Lab Mixer
  • Ready to use system comes complete with three FREE accessories!
  • Free accessories include: Boss head clamp, propeller, and stand
  • Exceptional value
from $599.00 More details
Cole-Parmer Digital Reversing Mixer with Remote Controller
Laboratory Mixer with Remote Control
  • Unique, dual-keypad operation allows for complete mixer adjustment up to six feet away!
  • Remote controller
  • Reversing and timed cycles
from $1200.00 More details
Laboratory Mixer Accessories
Lab Mixer Accessories
  • Fulfill all of your mixing needs with our array of accessories
  • Choose from a wide assortment of accessories from propellers to stands
  • Propellers available in a variety of materials including stainless steel and PTFE
from $13.00 More details
Laboratory Mixers and Accessories
Cole-Parmer offers an extensive line of overhead laboratory mixers and accessories to suit mixing requirements from lab to scale-up. Our full line of laboratory mixers includes analog, digital, and RS232 compatible lab mixers. See the table below for more detailed information on our entire line of lab mixers.
When considering your laboratory mixing needs, consideration has to be given to a wide range of factors including:
  • Container capacity
  • Liquid viscosity
  • Torque
  • Horsepower (hp)
  • Rotational speed (rpm) and diameter of mixing propeller
  • Duty cycle
  • Power supply
For more information on these variables, check out our Laboratory Mixer Selection Guide for definitions and requirements. Our Parametric Search for Laboratory Mixers allows you to easily search our entire selection of mixers and returns those that fit your requirments. In addition to these requirements, the position of an overhead mixer in a laboratory vessel will create different mixing patterns and lead to different results. For more information on positioning a laboratory mixer in a vessel, see Positioning a Mixer in a Cylindrical Vessel.
View our online catalog to see our full selection of products or contact our Application Specialists to help you find the right product for your needs.
Laboratory Mixer Selection Table
A laboratory mixer allows for mixing of smaller quantities of material, typically up to 100 gallons (380 liters). Lab mixers are capable of handling solutions with viscosities up to 150,000 cps, which is dependent on the torque, horsepower, and speed of the mixer. Laboratory mixers are also available with the following options:
  • Speeds up to 10,000 rpm
  • Through-shaft design for easy shaft and propeller adjustment
  • Digital display of speed, torque, and timer functions
  • Sample light for easy viewing during low light conditions
  • Remote controller for easy adjustment from up to six feet away
  • RS232 connectivity for easy data collection
Image Type Applications Features Specifications
Cole-Parmer® Ultra-Compact Digital Mixer General, non-viscous mixing
  • Digital display
  • Mixing speeds up to 2500rpm
  • Complete system includes mixer, stand, shaft, and two propellers
  • Speed: 60 to 2500 rpm
  • Motor: 1/100 hp Brushless DC
  • Maximum mixing capacity: 2 liters
  • Display: Digital
  • Cole-Parmer Stir-Pak® General Purpose Mixer Routine, light-duty mixing of water-like substances
  • Analog control
  • Mixing speeds up to 10,000 rpm
  • Compact size
  • Speed: 500 to 10,000 rpm
  • Motor: 1/15 hp fuse protected
  • Display: Analog
  • NEW Cole-Parmer Stir-Pak Dual-Shaft Mixer General mixing requiring either high-speed, low viscosity mixing or low-speed, high viscosity mixing
  • Two motor shafts for more versatility
  • Solid-state controller for precise speed control
  • Totally-enclosed, non-vented motor
  • Speed(direct mixing): 50 to 500 rpm
  • Speed(gear shaft): 3 to 250 rpm
  • Motor: 1/50 hp TENV dual-shaft
  • Display: Analog
  • NEW Cole-Parmer Heavy-Duty Stir-Pak Mixers System flexibility for your mixing needs
  • Mixing heads provide speeds of 2 to 180 rpm up to 60 to 6000 rpm
  • Separate analog controller with 6-ft cord
  • Available in high torque, low speed or high speed, low torque versions
  • Speed(high-torque, low speed):
    2-180 or 9-900 rpm
  • Speed (high-speed, low torque):
    23-2300 or 60-6000 rpm
  • Motor: 1/15 to 1/10 hp TENV
  • Display: Analog
  • NEW Cole-Parmer Servodyne Mixers Digital mixing with computer interface for optional data collection
  • Mixing heads provide speeds of 3 to 180 rpm up to 150 to 6000 rpm
  • Separate digital controller with 6-ft cord
  • Available in high torque, low speed or high speed, low torque versions
  • Optional computer software allows for easy monitoring and control of your mixer
  • Speed(high-torque, low speed):
    3-180 or 20-900 rpm
  • Speed (high-speed, low torque):
    60-2300 or 150-6000 rpm
  • Motor: 1/15 to 1/10 hp TENV
  • Display: Digital
  • Cole-Parmer Digital Reversing Mixer Timed and reversed mixing for samples up to 20,000 cps
  • Timed mixing cycles for automatic reversing or shut-off
  • Reversing motor for better mixing
  • Intregrated light for better sample viewing
  • Speed: 40 to 2010 rpm
  • Motor: 1/10 hp Brushless DC
  • Maximum mixing capacity: 25 liters
  • Display: Digital
  • Cole-Parmer Digital Reversing Mixer with Remote Controller Timed and reversed mixing for samples up to 20,000 cps with removable remote controller for operation up to 6-feet away
  • Removable remote controller for operation up to 6-ft away
  • Lock-out feature prevents inadvertent changes
  • Timed mixing cycles for automatic reversing or shut-off
  • Reversing motor for better mixing
  • Intregrated light for better sample viewing
  • Speed: 40-2000 or 80-4000 rpm
  • Motor: 1/10 hp Brushless DC
  • Maximum mixing capacity: 25 liters
  • Display: Digital
  • Talboys Low-Cost Laboratory Mixers Light- to heavy-duty mixing with multiple configurations
  • Continuous duty mixers with overload protection
  • Varying mixer sizes provide different speeds and torque
  • Different configurations prvide flexibility
  • Speed: Varying ranges from 50 to 7500 rpm
  • Motor: 1/75 to 1/4 hp
  • Display: Analog
  • Economical Constant-Torque Mixers Varying speed and torque ranges for mixing low- to high-viscosity solutions
  • Maintain constant torque throughout speed range
  • Totally enclosed non-ventilated (TENV) motors seal out dust and fumes
  • Integrated control box for on/off and speed control
  • Speed: Varying ranges from 0 to 6000 rpm
  • Motor: 1/15 to 1/8 hp
  • Display: Analog
  • Compact Digital Lab Mixer Accurate and repeatable mixing for viscosities up to 15,000 cps
  • Integrated sample light illuminates solution being mixed
  • Mixes up to 25 liters of solution
  • Digital control panel makes it easy to reproduce mixing parameters
  • Speed: 40 to 2000 rpm
  • Motor: 1/15 hp Brushless DC
  • Maximum mixing capacity: 25 liters
  • Display: Digital
  • Heidolph Laboratory Mixers Varying speed and torque ranges for mixing low- to high-viscosity solutions
  • Stepless speed control
  • Provides speeds up to 2700 rpm
  • Capable of mixing low- to high-viscosity solutions
  • Speed: 12 to 2000 rpm
  • Motor: 1/40 to 1/8 hp Brushless DC
  • Display: Analog or Digital
  • GK Heller® Dual-Shaft Mixer Mixing of low- to high-viscosity solutions
  • Dual-shafts provide both low and high speed mixing
  • Maintains constant speed regardless of changes in torque
  • TENV motor provides pulse-less mixing
  • Speed: 0 to 4000 rpm
  • Motor: 1/40 hp TENV
  • Display: Analog
  • GK Heller Mixers with Digital Speed & Torque Readout Mixing of low- to high-viscosity solutions, ideal for polymer chemistry
  • Maintains constant speed and torque
  • Electronically control and monitor speed and torque
  • Easy gathering of information for viscosity profiles
  • Speed: 2 to 2500 rpm
  • Motor: 1/18 to 1/4 hp TENV
  • Display: Digital
  • IKA Laboratory Mixers and Accessories Provide mixing of solutions with viscosities up to 150,000 cps
  • Mixers range from general purpose to high-viscosity mixers for process scale-up
  • Choose from a selection of analog or digital units
  • Accessories include stands, clamps, and impellers
  • Speed: Varies depending on model
  • Motor: Varies depending on model
  • Display: Analog or Digital
  • Real-Torque Digital Mixers Constant speed as viscosity of your solution changes
  • Constant performance under changing demands
  • Chuck guard protects you from spinning chuck
  • Through-shaft motor for easy depth changes of propeller
  • Speed: 20 to 6000 rpm
  • Motor: TENV
  • Display: Digital
  • Lightnin® Lab Master Mixers General purpose and computer compatible models to fit a variety of needs
  • Ideal for pilot studies of various processes
  • General purpose mixers offer either constant speed or remote controlled units
  • Digital LabMaster units provide repeatable data entry and ensure accurate scale-up
  • Speed: Varies depending on model
  • Motor: Varies depending on model
  • Display: Analog or Digital
  • Air Powered Laboratory Mixers Provides mixing where electricity is unavailable or restricted
  • Speeds available up to 10,000 rpm
  • Direct and gear drive mixers available
  • Low and high torque models available for varying viscosities
  • Speed:
    Varies depending on model and air pressure
  • Motor: Air drive
  • Display: None
  • Cole-Parmer
    625 East Bunker Court
    Vernon Hills, Illinois 60061
    Phone: 1-800-323-4340
    Fax: 1-847-247-2929
    E-Mail: info@Coleparmer.com
    Copyright © 2010
    Cole-Parmer Sites  Feedback
    Cole-Parmer:
    United States
    Canada
    China
    United Kingdom
    India
    McAfee Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams