Do you ever feel stuck at the lab monitoring samples and even missed a few after-work activities, because you feared if you walked away, you might lose valuable samples? If you are someone who wants to be connected, here’s a perfect solution for you at an affordable price.
Traceable® data loggers equipped with TraceableLive® wireless technology gives you the freedom to go where you need to go and stay connected. This technology is ideal for those who want to monitor critical environments and get alerts anywhere. Plus, the included NIST-traceable calibration at a noncalibrated price provides value, accuracy and peace-of-mind.
How the data loggers work
Traceable data loggers equipped with TraceableLIVE provide a simple, efficient and reliable way to ensure critical samples are not compromised due to parameter variations. The data loggers are also equipped with NIST-Traceable calibration, which means reliable measurements right out of the box saving both time and money. The following four Digi-Sense data loggers have these features:
- Traceable Temperature/Humidity Data Loggers with a range of 32 to 131°F (0 to 55°C)
- Traceable Ultra-Low Temperature Data Loggers to measure down to –130°F (–90°C)
- Traceable Multiparameter Data Loggers with TraceableLive to measure temperature, humidity, CO₂, or barometric pressure levels
- Traceable Data Logging Refrigerator/Freezer Thermometers with the choice of four different temperature probe styles
TraceableLive securely connects to data via Wi-Fi on a smartphone, tablet or PC. The cloud-based interface requires no additional software, and connecting is simple and easy with a low-cost TraceableLIVE subscription. Once setup is complete, workers can view conditions in realtime; set alarm parameters; view data logging history; generate data reports in real time; get email, text and push notifications; and receive alerts for temperature alarm, connectivity interruption, and low battery.
Learn more about TraceableLive
Be the first to comment on "Monitor Critical Samples with Traceable Data Loggers"