New Jenway 74 Series Spectrophotometers Feature CPLive™ Connectivity
spectrophotometer
Jenway 7415 Scanning Spectrophotometers with CPLive Connectivity
Spectrophotometers with Enhanced Connectivity
The new Jenway 74 Series Visible and UV/Visible Scanning Spectrophotometers with CPLive Connectivity gives you an easier way to experience spectroscopy. Safely and securely upload and archive data to the cloud, manage multiple devices at one time with the CPLive App, share data with colleagues, and access unlimited data storage. The Jenway 74 Series spectrophotometers provide accurate and reliable results ideal for a variety of applications from teaching, to industrial applications, to routine sample analysis in quality control environments.
Additional Features
You will find these spectrophotometers easy to use with a high-definition, color touchscreen, and intuitive user interface. The touchscreen interface is fast and responsive even when wearing gloves. With these spectrophotometers, you also have 10GB of onboard storage for methods and results, multiple USB ports for data storage and printer connectivity, multi-language options, and versatility with an extensive range of optional accessories.
Three models available
Model 7410 uses a tungsten halogen lamp for measurements in the visible spectrum from 320 to 1000 nm. Model 7415 uses a press-to-read xenon lamp to extend measurements into the UV spectrum down to 198 nm. The xenon lamp has a 3-year warranty. Model 7415 Nano is a micro-volume spectrophotometer.
To learn more about the Jenway 74 Series, go to ColeParmer.com/Jenway or call our product experts at 1-800-323-4340.
Browse our complete line of spectrophotometers.
Be the first to comment on "New Jenway 74 Series Spectrophotometers Feature CPLive™ Connectivity"