Nearly every lab needs a good supply of sample containers. Which ones to choose? From bottles to beakers to vials, a variety of labware is available. Yet not all will meet your specific needs.
Chemicals can affect the weight, strength, color, dimensions, flexibility, and surface appearance of your labware. They can soften and swell plastic and cause the permeation of solvent through the plastic. Temperature, pressure, internal or external stresses (such as centrifugation), can also impact chemical resistance.
Make sure the containers you use are chemically compatible with the chemicals you are placing in them.
Graduated polypropylene (PP) containers tolerate strong acids, alkalies, and most organic solvents. In addition to offering chemical resistance, these containers are:
- Translucent
- Unbreakable
- Available in sterile or nonsterile versions
These containers come with identification labels and color-coded HDPE caps with full-turn screw thread to provide a leakproof seal. The sterile containers are individually bagged.
See the graduated polypropylene (PP) containers and other options in labware.
Be the first to comment on "3 Reasons to Use Sterile Graduated Sample Containers"