4 New Time-Saving Products for Environmental Labs
Disposable BOD bottles
From wet chemistry to metals analysis to microbiology, get the products that will make your lab more efficient:BODBotttles
  1. Save time! No more cleaning BOD bottles. Disposable BOD bottles provide huge labor savings and improved quality. Lightweight unbreakable and recyclable disposable bottles offer safety as well. Bottles are formed from PET resin—the most easily recycled plastic available. Simply use once and toss into the recycling bin. An amorphous carbon coating keeps oxygen from diffusing into or out of your sample.
  2. Simplify and improve metals prep. HotBlocks are the answer for laboratories seeking faster, cleaner, more cost-effective methods of performing metals digestions.
  3. Save time and improve accuracy with colony counters, The Flash & Grow is an automated system, ideal for laboratories engaged in drug development and production, food and milk testing, environmental monitoring and public health, with a variety of budget possibilities. The Flash & Grow lets you create different configurations optimized for each application.
  4. Meet the demands of the Toxicity Characteristic Leaching Procedure. The MaxFil hazardous waste filtration system makes complying with Method 1311 easy and efficient. No splitting of the sample extract is necessary because the 2200mL capacity allows you to load the entire sample before filtration begins. This not only saves time but also reduces the supplies necessary to prepare a sample. Using a quick connect fitting instead of a cumbersome hose-barb fitting achieves pressurization. This quick-connect requires no clamping and, unlike a barbed fitting, it cannot “blow off” during filtration.
See even more.
Be the first to comment on "4 New Time-Saving Products for Environmental Labs"