Lobe pumps transport many solids and liquids, making them the choice for food producers, pharmaceutical manufacturers, and biotechnologists. The Sanitary Washdown Rotary Lobe Pump is one such pump, with other clear advantages including:
- Conforms to 3A sanitary standards
- Low maintenance
- High-pressure capability with heavy-duty shafts
- Reduced risk of shed particles in fluid with use of noncontacting rotors
- Reduced noise and cavitation for a safer, more comfortable work environment
The rugged design of this pump also features bi-wing rotors that require no timing adjustments—and provides a high volumetric efficiency. Front mounted shaft seals are fully accessible from the front of the pump for easy replacement without disturbing piping in the process line.
The Sanitary Washdown Rotary Lobe Pump is approved to EHEDG (European Hygienic Equipment Design Group) and CIP/SIP standards.
Excellent post. The sanitary washdown are useful for food producers, pharmaceutical manufacturers, and biotechnologists to keep their products hygienic.