5 Types of Motor Enclosures
Single-Phase TEFC Rigid-Base Motor
Single-Phase TEFC Rigid-Base Motor
When you are choosing a motor enclosure, you may be wondering what all the acronyms in the description mean. TENV? WDN? Here’s a list of the enclosure types and what each indicates in terms of use:
  1. Open Drip-Proof (ODP) Motors are equipped with open enclosures. These motors are suitable for indoor use and clean atmospheres. Ventilator openings are designed to prevent liquids and solids from entering the machine from an angle of 0 to 15°.
  2. Totally Enclosed, Fan-Cooled (TEFC) Motors have an external fan which circulates outside air to the motor.
  3. Totally Enclosed, Nonvented (TENV) Motors do not have air openings. While air does not freely move between the inside and outside, the motor’s construction is not air- or liquid-tight.
  4. Washdown (WDN) Enclosures work well in environments in which regular cleaning or sanitizing is necessary.
  5. Explosion-Proof (XPRF) Motors are completely enclosed to withstand an internal explosion of gas or vapor—the motor frame will not rupture or burst. Use these motors for safe operation in hazardous environments.
3 Comments on "5 Types of Motor Enclosures"
  1. Dear sir ,
    Kindly provide quotation for below item and provide delivery time, gross weight
    GEARED MOTOR, 1.5 KW, GEAR RATIO: 1/15, MOTOR
    FRAME SIZE: 90, SUPPLY: 380V A.C., 60 HZ, 1735 RPM,
    4 POLE, MODEL: FHB ; TEFC,
    MAKE: HICO (S.KOREA)
    QTY—–05 pcs
    Your immediate actin shall be highly appreciated
    • Hello:
      Thank you for your inquiry. I will send this to our customer service area. In the meantime, you can call 1-800-323-4340 for immediate information.
  2. Masereka Hannington | February 12, 2020 at 9:57 am | Reply
    SO INTERESTING LEARNING INFORMATION,Please keep posting.