If you live in the Eastern or Midwestern United States, there’s no need to guess how cold it is outside this winter ―the new H-B Instrument Enviro-Safe® Nonhazardous Glass Thermometers available from Cole-Parmer will confirm that.
Cole-Parmer is proud to present these new Earth-friendly “green” thermometers certified as 100% nontoxic, nonhazardous, and biodegradable. These accurate and safe thermometers offer the same accuracy as mercury thermometers without the hazards. They are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications where the presence of mercury or other hazardous liquids is not desired.
All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as nontoxic, nonhazardous, and fully biodegradable. In case of breakage, the liquid can be cleaned up easily with soap and water. These thermometers also have recyclable packaging, an antiroll fitting, individual serial numbers, and a statement of accuracy.
So, what’s so bad about mercury thermometers, anyway?
Mercury thermometers are harmful to both the environment and your personal health. In fact, Mercury is toxic to humans and wildlife. If a mercury thermometer breaks and is not properly disposed of, tiny droplets of mercury can evaporate within the environment over time. When mercury vapor enters the bloodstream through inhalation, brain damage, spinal cord, kidney, and liver damage may occur in time.
Additionally, improperly disposing of mercury from thermometers can contaminate lakes and streams. Mercury may accumulate in the bodies of fish and other wildlife and, through natural processes, converts this mercury into methyl mercury, which is harmful to humans when consumed.
Be the first to comment on "“Baby, it’s Cold Outside”"