Shield Your Eyes! How to Protect Your Sight
With a nod to January as National Eye Care month, we are devoting today’s blog to solutions that protect your eyes. If you are at risk of accidental exposure to corrosive or caustic materials, you may need to have an eyewash station nearby. Several options are available. Here are a few of our favorites:
Fend-all Personal Eyewash Wall Stations
Emergency eyewash stations hold one or two bottles of flushing solution and affix easily to any wall with peel-off self-adhesive backing or mounting screws (included). Eyewash bottles have smaller openings to provide extended flow. Sealed solution bottles have a 36-month shelf life. Expiration date is printed on each bottle.
Portable Eyewash                                              
Goes anywhere to provide onsite emergency relief for eyes and face. Yellow, reinforced thermoplastic drench hose (8-ft) is designed to supply supplemental water to face and body. Stainless steel tank features tire filler valve and 0-100 psi air gauge. Pressure relief valve prevents accidental over-pressurization.
Bradley® On-Site Portable Gravity-Fed Eyewash
The highly visible Bradley On-Site transparent polycarbonate yellow tank lets you view when it’s time to refill or clean. Refillable—mix one 8 oz. bottle of water preservative with seven gallons of potable water to prevent growth in tank for up to six months. Solution can be tested monthly or when protocol dictates. The removable tank is inverted onto pedestal, piercing a liner. Water flow is activated when arm is pulled down.
 Disclaimer: Cole-Parmer products are not approved or intended for, and should not be used for medical, clinical, surgical or other patient-oriented applications.
2 Comments on "Shield Your Eyes! How to Protect Your Sight"
  1. Those are very effective eye care products. Eyewash solutions aside, the best way to protect your eyes would be to do your best to shield your eyes from irritants or at least minimize your exposure to them by wearing a full face mask or shield. Absent an eyewash solution, the best alternative is clean, running water. It can help reduce any damage done until help arrives.
  2. Eye wash station is the best way to protect our eyes. Thanks for the Post.