Easy-to-Use and Compact: Shimadzu UV/Visible Scanning Spectrophotometers
Shimadzu UV/Visible Scanning SpectrophotometerReduce the time it takes to get up to speed and receive results! The Shimadzu UV/Visible Scanning Spectrophotometers feature large displays to quickly read graphical results as well as screen prompts to easily guide users. They offer full-range scanning in an overall smaller size. Scan over a range of 190 to 1100 nm.
Shimadzu UV/Visible Scanning Spectrophotometers are available in several models. The UV mini-1240 offers functionality from simple concentration measurements to sophisticated quantitative calibration curves. The BioSpec-mini is preprogrammed for nucleic acid quantitation and protein assays and analysis. The UV-1800 supplies data input/output and PC control via USB connectivity.
Optional accessories for the UV mini-1240 model include DNA/Protein Program Pack Software, Multiwavelength Program Pack Software, Kinetics Program Pack Software, UV Data Manager Software, and Thermal Printer Kit.
Be the first to comment on "Easy-to-Use and Compact: Shimadzu UV/Visible Scanning Spectrophotometers"