Conductivity measures the electrolytes in a solution. When water is polluted, its conductivity increases. Measuring conductivity can indicate how much pollution is in a water sample. This is tested by the ability of the solution to conduct electric current. From the water we drink to the water we swim in, conductivity is a crucial measurement.
- How does temperature affect conductivity readings?
It depends on the solution you are measuring. The effect of temperature is greatest in low conductivity (or low ionic strength) solutions. In most cases, there will be a 2% increase per °C for most aqueous solutions. However, organics have very different temperature curves.
- How are conductivity and TDS related?
Salts, minerals, and dissolved gases contribute to conductivity, meaning conductivity indicates the amount of dissolved materials in a solution. TDS can give you a fairly accurate measurement when comparing the status of a single source (such as NaCL). Yet when comparing two different types of solutions it may be error-prone. Calibrate the meter using the same dissolved materials that are in the test solution.
- What is the difference between conductivity and salinity?
The probe is the same for both, but a salinity meter has a correction factor applied to the reading. The correction factor takes the conductivity reading and converts it to ppm of a specific salt (which varies by manufacturer).
- Are conductivity probes interchangeable with meters?
No. The cell constant may be different and the pin configurations are typically different. The type of thermistor used for temperature compensation is also different.
- How and when do I need to calibrate the probe?
Use a standard solution in the range of samples you are testing to calibrate the probe. Place the probe in the standard solution, condition, and rinse the probe in a second sample of the standard solution. Then use a third sample of standard solution to calibrate, and adjust the cell constant until the specified value is displayed. Recalibrate when you change ranges or if readings seem inaccurate.
View our selection of conductivity meters and conductivity calibration solutions.
Be the first to comment on "5 Frequently Asked Questions about Conductivity"