Economical New Glass Labware Offered with Quantity Discounts
Cole-Parmer® Erlenmeyer Glass Flasks, 100 mL, 12/pkg
Cole-Parmer® Erlenmeyer Glass Flasks, 100 mL, 12/pkg
Cole-Parmer introduces three new laboratory items crafted of durable glass yet offered at stock-up prices. The flasks, bottles, and beaker have reduced prices when purchased in package or case quantities of five or ten.*
New Cole-Parmer® Erlenmeyer Glass Flasks, 100 mL, 12/pkg
 Cole-Parmer® Erlenmeyer Glass Flasks, 100 mL, 12/pkg
Cole-Parmer® Erlenmeyer Glass Flasks, 100 mL, 12/pkg
These flasks provide a low-cost alternative to fit every budget. Designed with a narrow-mouth opening and beaded heavy-duty rim, the flasks have approximate graduations and marking spot in durable white enamel. The Class A borosilicate glass meets ASTM E 1404, DIN 12380 specifications.
New Cole-Parmer® Class A Media Glass Bottles, 100mL, 10/case
These affordable borosilicate bottles are excellent for storage, mixing, and sampling needs. Quickly identify contents by large marking spot and graduations. The bottles have a wide-mouth opening for easy pouring and no-liner, blue polypropylene cap and pouring ring. Sturdy borosilicate Class A glass meets ASTM E438, Type 1, Class A requirements.
New Cole-Parmer® Griffin Low-Form Glass Beaker, 10mL, 12/pk
Easy-to-read double-scale white graduations make these beakers handy in the lab (10-mL beaker does not have graduations). They withstand temperatures up to 752°F (400°C) for short-term exposure and 445°F (230°C) for long-term exposure. Heavy-duty Class A borosilicate beakers meet ASTM E960 specifications.
View our full selection of new products.        
*Discount is valid in the U.S. and Canada only.
Be the first to comment on "Economical New Glass Labware Offered with Quantity Discounts"