Heating torches can do more than solder metals; they perform heat sealing in a variety of manufacturing, construction, and jewelry-making applications. The microtorch is especially handy, with a pinpoint flame that is adjustable for wider targets.
Four ways to use heating torches include:
- Heat shrink tubing and specialty connectors
- Spot dry
- Heat wax
- Work with laboratory glassware and glass tubing
To do their job, these torches have flame temperatures that reach as high as 2500°F (1300°C). You can select table-top models for laboratory and industrial uses or the convenient handheld portable models for field and other applications. Soldering pens are also available.
View our selection of heating torches.
Be the first to comment on "4 Reasons to Use Heating Torches"