Need to measure several parameters simultaneously? A multiparameter meter is a worthwhile investment. Usually these meters test pH, conductivity, dissolved oxygen, temperature, turbidity, depth, barometric pressure, and/or salinity.
Rather than juggling a few different meters, the multiparameter meter gets the job done with one instrument. Plus, you’re not paying to purchase—or calibrate—multiple instruments.
Environmental specialists, lab technicians, and research scientists regularly use multiparameter water quality meters in their work. The simultaneous readings keep them from missing key data.
What can you expect from these meters? In addition to taking many types of measurements, they may include data management software for tracking data, a probe holder, and some include a multilanguage interface and carrying case.
Check out our full selection of various multiparameter meters.
Be the first to comment on "Multiparameter Meters Add Versatility"