4 Essential Products for Biopharma
Sterilized Sampling Bottle Assembly
Sterilized Sampling Bottle Assembly
Processes in the biopharma industry must be sterile, precise, and validated. With the specialized needs of biopharma, product selection is critical. Here are four that have resonated with users:
  • Safely transfer, sample, and store biopharmaceutical products and reagents with the Sterilized Sampling Bottle Assembly. Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections.
  • B-D Plastic Disposable Syringe, Luer-Lock Tip is a sterile, polypropylene syringe in an easy-to-open sealed package. The syringes can be autoclaved out of the package.
  • PressureMAT Pressure/Flow Monitor is ideal for tangential flow filtration, cross flow filtration, depth filtration, bioreactor monitoring, and chromatography. The single-use solution for monitoring process pressure!
  • Use Whatman Polycap TF PTFE Capsule Filter or PP filter capsules for high-efficiency filtration and nylon filter capsules for filtering aqueous solutions. Nylon membranes are hydrophilic and low in extractables; they include a prefilter to prolong the life of the membrane. All capsules feature sturdy PP housings with fused seals-no glues or adhesives to contaminate filtrate.
Be the first to comment on "4 Essential Products for Biopharma"