Remove Condensation from Your Lab
Cast Iron Float/Thermostatic Steam Trap
Cast Iron Float/Thermostatic Steam Trap
Steam traps discharge condensate to provide greater efficiency in your lab. Essentially steam traps operate as valves to prevent valuable steam from being wasted, yet remove the unnecessary condensate. Generally, steam traps are energy-efficient and fairly economical.
Select from these configurations:
  • Thermodynamic, for drip applications, superheated steam applications, steam main drain and tracer lines, process equipment, laundry and kitchen equipment, outdoor installations subject to freezing
  • Thermostatic, for process and tracing applications, heat exchangers, air heating coils, and more
  • Inverted bucket for industrial dryers, storage tanks, laundry equipment, and steam main drip stations
Air eliminators improve the circulation of pressurized liquids by eliminating air and other non-condensable gases. They are used for suction lines to pumps, cold water lines, condensate return lines, mixing tanks, cool water lines on air compressors, and water storage tanks.
See our selection of steam traps and air eliminators.
Be the first to comment on "Remove Condensation from Your Lab"