PCR programming is easier than ever—with the use of program templates. The new Cole-Parmer® StableTemp™ Thermal Cyclers include two- and three-step PCR, RT-PCR, chill, and ligation program templates. To enhance program versatility, the units also include incremental and decremental temperatures and step hold times.
The nongradient thermal cycler uses a Peltier heater system to produce uniform temperature.
Users can switch out thermal blocks for a quick change in testing protocol. Passwords protect saved setting configurations and temperature cycle programs to prevent tampering and accidents.
Personal/Compact Models have a small footprint to conserve bench space or fume hood use. These cyclers are designed for smaller sample batches, and feature faster heating and cooling rates than the full-size models. Full-Size Models are best suited for high throughput applications.
The thermal cyclers are designed to be workhorses in busy environments—multiple units can be daisy-chained together when used with user-supplied control software.
View all Thermal Cyclers.
Be the first to comment on "Program PCR Processes with New Thermal Cyclers"