You can now fill your tool belt with accurate, durable electrical test instruments without depleting your budget. The new line of Digi-Sense electrical handheld instruments, from Cole-Parmer, is a smart and economical alternative to higher-priced instruments on the market today. NIST-traceable calibration ensures instrument accuracy, adding value and peace of mind (included on select products). Electricians, engineers, technicians and more will appreciate these quality instruments that deliver consistent, reliable measurements.
Check out the new Digi-Sense electrical test instruments below:
Digital Multimeters
These handheld multimeters have special features designed for a wide number of applications. Each multimeter includes NIST-traceable calibration.
- Digi-Sense Three-in-One Digital Multimeter combines the most common voltage-current-resistance functions in a compact design at a very affordable price.
- Digi-Sense Compact Digital Multimeters feature noncontact voltage detection.
- Digi-Sense Industrial Digital Multimeters meet IP67 standards for dust and waterproof protection as well as CAT III 600 V and CAT III 1000 V ratings for safety.
Clamp Meters
These meters measure higher currents than a digital multitester, and feature a jaw designed to easily get into tight spaces. Each meter includes NIST-traceable calibration.
- Digi-Sense 400 A AC Clamp Meters have a compact design for testing in tough-to-reach places.
- Digi-Sense 1000 A AC Clamp Meters feature True RMS for improved accuracy.
Testers
Testers are ideal for checking safety conditions as well as front-line troubleshooting. These testers are not precalibrated.
- Digi-Sense AC/DC Multifunction Voltage Tester tests power outlets and switches for correct circuit wiring before resuming power.
- Digi-Sense 1kV Handheld Insulation Tester has live circuit detection to prevent testing if voltage is present.
- Digi-Sense Receptacle Testers are a fast and effortless way to test the safety of electrical socket outlets. Available for circuit and GFCI.
For more information about Digi-Sense electrical instruments, visit ColeParmer.com.
Dear Sir/Madam,
Good day.
I want to know if your company can supply your products to our Association.
However, we will like to see your company catalog/website to help select the Require items.
If you are interested then get back to enable us to proceed.
Waiting for your prompt response, and have a nice day.
In the meantime, I look forward to hearing from you by return of my mail and to developing a long-term business partnership with you soon.
Best Regards,
Nana Mensah
Hi Nana:
All of our products are on our coleparmer.com website. You can purchase what you need there! Thank you for reaching out.
Hi Nana:
We do not print catalogs but you can purhcase what you need from our website! Thank you for reaching out.