Cole-Parmer® Precalibrated Compact Digital Multimeters
Pocket sized—have on hand wherever you go
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
  • Easily select a function with a single rotary switch
  • Stay safe and prevent damage with overload protection
  • Increased durability with double-molded plastic housing
  • Tilt stand for easy hands-free viewing
  • CAT III-600 V safety rating
Versatile and simple-to-use digital multimeters safely measure common ranges of voltage, current, and resistance, and are packaged in a very convenient compact size. They feature diode testing, continuity check with audible buzzer, a measurement rate of 2 readings per second, overrange and low-battery indication, and auto power-off.
Basic model 20250-50 offers manual ranging and measures AC and DC voltage to 600 V, resistance with continuity and diode function, and DC current to 10 A. Plus a dedicated battery test function lets you check the quality of your 9 V and 1.5 V batteries.
Full-featured model 20250-51 also measures AC current to 10 A, capacitance, frequency, duty cycle, and temperature via a type K thermocouple probe. An autoranging feature automatically selects the optimal measuring range for maximum resolution. Plus you’ll work more efficiently with the data HOLD and relative functions.
ITEM(S) ON SALE
$41.12 - $52.80USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Ac Voltage Accuracy DC Voltage Accuracy Description
Availability Pricing
COMPARE
±1.2% ±0.8% Compact Digital Multimeter, Manual Ranging with NIST Traceable Calibration
$41.12USD / Each
$51.40USD / Each
COMPARE
±2.0% ±1.5% Compact Digital Multimeter, Auto Ranging with NIST Traceable Calibration
$52.80USD / Each
$66.00USD / Each