What are the types of stirrers, hot plates and stirring hot plates?
- Compact: Small, colorful units fit in any lab.
- Multiposition: An illuminating stirrer and a modular unit that offers remote immersible mixing.
- Multiposition/Immersible: Immersible units can be placed completely underwater for constant temperature operations. Ideal for use in incubators.
- Specialty: These can include modular units for flexibility and remote immersible mixing.
- Analog: Operate these manually by turning the dial. They typically are the most economical of all units and ideal for basic stirring and heating.
- Digital: Easy to use with the push of a button, digital units can provide greater accuracy and more features. Programmable models can be programmed directly from the front panel to control speed, temperature, and run time.
- Large Capacity: Larger than the typical sizes, these units can handle up to 150 liters.
- Explosion Proof: Ideal for operation in Class I, Group C and D environments.
- Heavy Duty: Made to withstand even the toughest environments.
Need more information?
Read our Selection Guide
Related Articles
5 Reasons to Use Stirring Hot Plates with Antimicrobial Coating
Lab Shakers and Stirrers: Movement for Successful Science
Be the first to comment on "A Guide to Stirrers, Hot Plates and Stirring Hot Plates"