New Compound Microscopes Focus on Clarity and Comfort
microscope terminology
Look into these compound microscopes ideal for both students and professionals.
A compound microscope consists of a group of lenses (called objectives) which focuses a real image of the object inside the microscope. A second group of lenses (called eyepieces) magnify this image and projects it on the retina of the eye. This compound optical system (objectives – eyepieces) together with other mechanical components are the basics of a modern microscope. The overall resolution and image quality of a microscope is determined by the quality of the optics and the light source.
New Cole-Parmer compound microscopes offer everything for guaranteed performance including advanced optics, high-quality materials and components, and ergonomics to meet today’s standards. All provide excellent clarity and resolution, are rugged and sturdy, and comfort while minimizing fatigue. A variety of models are available for uses in education, life sciences, veterinary offices and biomedical laboratories. The new compound microscope portfolio includes a variety of options.
Cole-Parmer MSU Series compound microscopes
Compact and designed for viewing small samples less than 7 micron. Available in four MSU Series with a variety of models.
objective lenses
MSU-100 and MSU-200 Series compound microscopes are entry-level models, easy to use and rugged—ideal for education. They can withstand extensive use and rough handling. High-quality, yet affordably priced, these microscopes match these demands due to the design and use of high-quality materials.
compound microscopes
MSU-400 and 600 Series compound microscopes are versatile and advanced microscopes with the finest optical components and smooth operation for effortless objective changes and optimal viewing—ideal for higher education and professional laboratories. Binocular and trinocular microscopes are available.
All microscopes are equipped with energy-efficient LED illumination, antibacterial protection, integrated X-Y mechanical stage, rotating 360-degrees head, Abbe condenser, ergonomics for the utmost comfort, and more.
How to select a compound microscope
If you’re unsure of which compound microscope to purchase, there are some factors to consider. Five key elements to look at are the purpose you’ll be using it for, the type of objective lenses, the amount of eyepieces, the total magnification, and the lighting. Investigate thoroughly before selecting a microscope or try out our selection tools.
Download the Microscope Selection Guide.
Related Articles
What is a Compound Microscope?
What are Objective Lenses on a Microscope?
What are Infinity Optics on a Microscope?
Be the first to comment on "New Compound Microscopes Focus on Clarity and Comfort"