Encore Tech Challenge: Using Sieves to Identify Mesh Range
TECHCHALLENGE[1]Pam runs Powder Inc’s Research and Development lab. She is looking for sieves to help identify the mesh range of their activated charcoal powder, as well as quantify (for quality reasons) the percentage that falls into each mesh size. They expect a range of 200 to 400 mesh. What would you suggest?
Sieves
A: The Cole-Parmer® sieve shaker for 3″ sieves and sieves
B: Standard 12″ testing sieves in a range from 200 to 400 mesh
C: Standard 8″ testing sieves in a range from 200 to 400 mesh
D: A Ro-Tap® sieve shaker with sieves
Answers:
A: The Cole-Parmer® sieve shaker for 3″ sieves and sieves. Correct! This economical shaker and selected sieves will separate into five mesh ranges. Plus, the uniform motion and digital timer provide reproducible results as compared to hand-sieving.
B. Standard 12″ Testing Sieves in a range from 200 to 400 mesh. Incorrect. Other more reproducible options are available.
C: Standard 8″ Testing Sieves in a range from 200 to 400 mesh. Incorrect. Other more reproducible options are available.
D: A Ro-Tap® sieve shaker with sieves. Incorrect. Although this would provide reproducible results in the mesh range required, there is a more economical option for Pam’s R&D testing needs.
Be the first to comment on "Encore Tech Challenge: Using Sieves to Identify Mesh Range"