Grab for Your Lab: Three New Feature-Rich Products
acrylic in line flowmeter with valve
Each of the new items below offers distinct advantages, from taking up less space to adding more efficient control.
Thermo-Scientific-VersaCool-Refrigerated-Circulating-BathThermo Scientific Versacool Refrigerated Circulating Bath
Innovative “headless” design creates more space for samples while maintaining a small footprint. By removing the control head and coil from the bath’s interior, you gain a large working area.Energy efficient and smart—it automatically detects appropriate frequency and voltage to work worldwide.
Denali Thermoelectric Chillers
Denali-M1-Recirculator-Chiller-by-Solid-State-Cooling-SystemsQuiet, vibration-free temperature control for cooling diode lasers, CCD cameras, and lab instruments. Smaller in size than comparable air-cooled recirculating chillers to save space. Offer precise digital PID control from 0° to 45°C.
Cole-Parmer® Valved Acrylic Flowmeters for Water and Gases
Gain easy control of your media with variable-area flowmeters offering direct reading scales for water, argon, oxygen, carbon dioxide, nitrogen, or helium. Multi-turn valve is included, and dual scales display flow rates in both metric and English units. Yellow backing enhances readability.
Be the first to comment on "Grab for Your Lab: Three New Feature-Rich Products"