Whether in industrial settings, manufacturing facilities, or as a matter of lab safety, gas monitors help protect workers by assessing their exposure to toxic and combustible gases. Air monitoring equipment is available in different varieties: an assessment must be conducted of potential hazards and then appropriate equipment chosen for use.
Here’s a description of various types of gas monitors:
Gas Detection Badges—there are two types: badges with a color change indicating the presence of the gas in question; and badges that are sent to a lab after use for analysis. Badges with color change indicators are designed for a single chemical, but have the advantage of showing immediate results. The badges sent in for analysis can be used to sample a broader array of chemicals with higher accuracy at lower levels of concentration.
Gas Detection Tubes—there are two types: diffusion tubes and detector tubes that use a pump. Diffusion tubes allow gas to enter the tube through diffusion, react with the reagent, and produce a color change with exposure levels read from markings on the tube. These are used for time-weight average (TWA) calculations. Detector tubes are used with a hand pump for quick sampling of an area vs. longer term measurements for an individual worker.
Single- and Multi-Gas Detectors—Can be worn by a worker or handheld, giving an immediate reading of gas concentration. Visible and/or audible alarms alert you to high toxic gas concentrations, or low oxygen levels. Some models have data logging capabilities for later download to a computer for record-keeping purposes.
Photo-ionization detectors (PID) detect volatile organic compounds (VOCs). They are sensitive to a broad band of VOCs eliminating the need for multiple sensors. For best results, they should be calibrated using the target gas.
Be the first to comment on "Industrial and Lab Safety: Monitoring Exposure to Toxic and Combustible Gases"