Lab Equipment for Genomic Research
Genomics is at the forefront of research, with the potential of genetic testing and personalized medicine, and the lab equipment described below populates the labs of genomic researchers. From linking enzyme deficiency with faulty DNA replication and cancer to identifying a new metabolic disorder caused by faulty gene expression, genomics researchers continue to make strides in this scientific frontier.
Equipment List for the Genomics Laboratory:
Centrifugal Concentrators for use with genomics including centrifugal filter units and concentrators
Cloning Products including cloning discs for genomics research and other life sciences
Electrophoresis Equipment includes horizontal and vertical systems, power supplies, reagents, blotting systems, and gel dryers
Hybridization Incubators for genomics include Minidizer, rotisserie, and shaking styles
Imaging Products include autoradiography film, BioDoc-It, DigiDoc-It, MultiDoc-It, and PhotoDoc-It systems
Electroporation Products from electroporators, buffers, cuvettes, to microslides for use with genomic research and studies
Thermal Cyclers also referred to as PCR machines and DNA amplifiers feature the ability to run numerous PCR samples simultaneously. Some units incorporate a thermal gradient function allowing different temperatures to be applied in various parts of the block
Polymerase Chain Reaction (PCR) Chambers from UV, UV Class 100, and other PCR stations reduce airborne contamination
Additional PCR Equipment from chilling/heating/mixing blocks, microcentrifuges, spectrophotometers, and thermal mixers
PCR Accessories include cryogenics, PCR plates, pipettors and tips, sealing films, and vacuum pumps
UV Crosslinkers or systems for UV crosslinking of DNA or RNA to membranes or gene apping for creating cleavage-inhibiting thymine dimmers
Disclaimer: Cole-Parmer products are not approved or intended for, and should not be used for, medical, clinical, surgical, or other patient-oriented applications.
Be the first to comment on "Lab Equipment for Genomic Research"