Laboratory Supplies to Help You Spring Clean Year Round
lab cleaning
Keep your lab and cleanroom efficient and maintain valuable equipment.
Cleaning products keep equipment running at peak efficiency, reduce the risk of contamination, and ensure the health and safety of personnel – a necessity for every lab and cleanroom facility,
Designating specific times of year to reorder supplies will also save your budget dollars, as you can buy in bulk and ensure you have what you need before it unexpectedly runs out. Here are a few customer favorites:
Cole-Parmer® Essentials DRNase Free™ Surface Decontaminant
Instantly eliminate DNase and RNase from labware and surfaces. The DRNase Free reagent surface decontaminant is ideal for items and surfaces used in PCR that cannot be autoclaved. Just spray and rinse.
Cole-Parmer® Essentials Micro-90 Cleaning Solution
Removes most biological material. Micro-90 is a mild, yet powerful, free-rinsing cleaner for use in critical cleaning and general purpose applications.
Cole-Parmer® Essentials Dry Clean-Wipes
Dry wipes can be used dry or can be used with any liquid. Simply add eight ounces of any fluid to the wipe container.
clean wipesCole-Parmer Essentials Clean Wipes
Premoistened wipes are available either with a blend of pure, reagent-grade 70% isopropyl alcohol and 30% reagent-grade deionized water, or 100% pure reagent-grade deionized water.
View all laboratory cleaning supplies.
Be the first to comment on "Laboratory Supplies to Help You Spring Clean Year Round"