Lab Equipment: A List of Microscope Terms
Microscope terms to help you understand how a microscope works. Abbe condenser—consists of one or two lenses with an additional specially designed lens mounted below the stage. An aperture controls both the amount of light…