Top Toploader Electronic Balances from Cole-Parmer
electronic balance
Get the best performance with these electronic balances.
Get best-performing features while still remaining budget savvy with these electronic balances. The expanded line of economical balances from the trusted Cole-Parmer brand include:
Cole-Parmer® PB-300 Series Portable Toploading Balance
This PB-300 series of toploading electronic balances features an ALIBI-compliant onboard memory. Ideally suited to regulated industries requiring data integrity and auditing capabilities, the ALIBI function prevents data modification and deletion. Printing and streaming of GLP-compliant weight reports are also available for industries subject to audit. Included data collection software allows you to export logged data for further analysis. USB A and B ports support device and PC connection, and RS-232 provides printer or PC connection. Balances may run on mains power, or use the built-in rechargeable battery where portability is necessary.
Cole-Parmer® PB-350 Series High-Capacity Portable Toploading Balances
This PB-350 series of electronic balances feature included data collection software allows you to export logged data for further analysis, and RS-232 interface provides printer or PC connection. Balances may run on mains power, or use the built-in rechargeable battery where portability is necessary.https://www.coleparmer.com/p/cole-parmer-pb-350-series-high-capacity-portable-toploading-balances/72691
Cole-Parmer® TB-800 Series Precision Touch-Screen Toploading Balances
The Cole-Parmer TB-800 series of precision toploading electronic balances offer advanced weighing at an affordable price! The balances feature several databases for recording products, users, packaging, customers, formulations, formulation reports, ambient conditions, and weighing. Only users with appropriate access can add new records, export data, import data, delete records, and print saved data. Two USB, two RS-232 interfaces, Ethernet, and Wi-Fi† port offer external connection to a computer or printer for GLP/GMP-compliant output. The autotest GLP function provides an enhanced balance summary to aid in assessing operation and diagnosing reasons for occurrence of errors in weighing.
See the complete line of toploading balances
Related Articles
The Laboratory Balance: Toploading vs Analytical
How to Troubleshoot Laboratory Balances
Be the first to comment on "Top Toploader Electronic Balances from Cole-Parmer"