Level Control: How do Electro-Optic Point-Level Sensors Work?
Electro-optic point-level sensors detect liquids regardless of conductivity, color, or specific gravity. These sensor types can be used for oil, water, and wastewater leak detection, food and beverage leak detection, semiconductor manufacturing, pharmaceuticals, chemical processing,…