Gas mass flowmeters only work with dry gasses. Normally the gasses are pure as well. If the gas contains too much moisture, an errant reading will occur. In this case a gas dryer is needed.
What else may be happening when your gas mass flowmeter is not operating properly? Here are some possibilities:
Problem: The reading is too high
Solution: Known as “swamping,” this happens when the flow exceeds 10% beyond the maximum-rated capacity. Lower the flow and shut system down completely to restore performance.
Problem: No display/power
Solutions: Check to make sure all cables are connected correctly and the power supply has been correctly selected according to requirements. When several meters are used on a power supply, an appropriate current rating should be selected. Possibly check for a blown fuse.
Problem: Incorrect reading
Solutions:
- Check for any leaks in the installation plumbing.
- Ensure the connector pinouts match properly. When interchanging with other manufacturers’ equipment, cables and connectors must be carefully wired for correct pin configurations.
- In case of moisture, allow moisture to dry out prior to continuing to use. Verify that the pressure differential across the instrument is sufficient.
- Clean/flush filter
- Adjust valve if required.
Be the first to comment on "Troubleshooting Gas Mass Flowmeters"