Protect Your Lab with Biosafety Cabinets
Purifier Cell Logic+, Class II A2 Biosafety Cabinet
Purifier Cell Logic+, Class II A2 Biosafety Cabinet
When you’re working with pathogens or other potentially hazardous material, it helps to have protection. Biosafety cabinets limit your exposure. These workspaces also keep your samples and processes from contamination while containing biological agents within the cabinet.
Ventilation of air volume within the cabinet determines its classification. Biosafety cabinets are designated as Class I, Class II, or Class III. For worker protection that does not protect samples, Class I is the first option. Class II (Type A and B) protects lab workers, samples, and the external environment. Class III is the most rigorous level of protection, with a gas-tight, enclosed structure for high risk biological agents. This level protects personnel, samples, and the external environment.
Some biosafety cabinets may have special features such as the ability for a microscope to be integrated into the cabinet. Another feature is a temperature-controlled area. Others are designed for chemical safety. Make sure the cabinet you choose has the capabilities and features to meet both your short term and long-term laboratory uses.
The CDC advises that the biosafety cabinet must be combined with “the appropriate practices, procedures and other administrative controls to further reduce the risk of exposure to potentially infectious microorganisms.”
For more information on classification, go to NSF International. The CDC also provides background on biosafety cabinets and classification. View our selection of biosafety cabinets or contact our technical experts if you have questions.
Be the first to comment on "Protect Your Lab with Biosafety Cabinets"